This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NKX6-2 SIRNA Assays ›
Gene Symbol:
Nkx6-2
Gene Name:
NK6 homeobox 2
Gene Aliases:
Gtx, Nkx6.2
Chromosome Location:
Chr.7: 139579376 - 139582797 on Build GRCm38
UniGene ID:
Mm.28308
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s201432

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Nkx6-2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183248.3 NP_899071.2 3 898
GenBank mRNA AK008173.1 - 3 899
BY718566.1 - 3 490
L08074.1 - 3 836

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001107558.

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
negative regulation of cell fate commitment
positive regulation of cell fate commitment
regulation of transcription from RNA polymerase II promoter involved in spinal cord motor neuron fate specification
regulation of transcription from RNA polymerase II promoter involved in ventral spinal cord interneuron specification
central nervous system myelination
cell differentiation
endocrine pancreas development
negative regulation of glial cell differentiation
positive regulation of glial cell differentiation
oligodendrocyte differentiation
neuromuscular process controlling balance
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm