This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CNIH1 SIRNA Assays ›
Gene Symbol:
Cnih1
Gene Name:
cornichon homolog 1 (Drosophila)
Gene Aliases:
0610007J15, CNIH-1, Cnih
Chromosome Location:
Chr.14: 46775567 - 46788888 on Build GRCm38
UniGene ID:
Mm.3261
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID s201176

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cnih1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009919.2 NP_034049.2 4 413
GenBank mRNA AF022811.1 - 4 404
AK002293.1 - 4 381
AK011200.1 - 4 366
AK152432.1 - 4 364
AK154479.1 - 4 364
AK168154.1 - 4 364
BC034868.1 - 4 373 2938052
DV653213.1 - 4 467

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001106029, BC158747, FQ222749.

Gene Ontology Categories:

Function(s) Process(es)

transport
biological_process
vesicle-mediated transport
intracellular signal transduction
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0