This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other UBE2H SIRNA Assays ›
Gene Symbol:
UBE2H
Gene Name:
ubiquitin conjugating enzyme E2 H
Gene Aliases:
E2-20K, GID3, UBC8, UBCH, UBCH2
Chromosome Location:
Chr.7: 129830732 - 129952960 on Build GRCh38
UniGene ID:
Hs.643548
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s197382

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: UBE2H
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001202498.1 NP_001189427.1 5, 6 310
NM_003344.3 NP_003335.1 5, 6 706
GenBank mRNA AB451490.1 - 5, 6 301
AK316590.1 - 5, 6 289
BC006277.2 - 5, 6 618 3957107
BI088146.1 - 5, 6 310
BT006756.1 - 5, 6 301
KJ534959.1 - 5, 6 301
Z29328.1 - 5, 6 354
Z29330.1 - 5, 6 354

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001172127, CB739657.
This siRNA also targets the following transcripts in Mouse: NM_001169577, NM_009459, BC008517, AK131688, U19854.

Gene Ontology Categories:

Function(s) Process(es)

ubiquitin-dependent protein catabolic process
proteasome-mediated ubiquitin-dependent protein catabolic process
protein K48-linked ubiquitination
protein K11-linked ubiquitination
ubiquitin-protein transferase activity
protein binding
ATP binding
ubiquitin protein ligase binding
ubiquitin protein ligase activity
ubiquitin conjugating enzyme activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header