This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ARPC2 SIRNA Assays ›
Gene Symbol:
ARPC2
Gene Name:
actin related protein 2/3 complex subunit 2
Gene Aliases:
ARC34, PNAS-139, PRO2446, p34-Arc
Chromosome Location:
Chr.2: 218217094 - 218254348 on Build GRCh38
UniGene ID:
Hs.529303
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s19670

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 357.00
Your Price
Online offer: 296.31
296.3
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ARPC2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005731.3 NP_005722.1 5 400
NM_152862.2 NP_690601.1 6 495
XM_011510471.1 XP_011508773.1 2 677
XM_017003113.1 XP_016858602.1 5 369
XM_017003114.1 XP_016858603.1 6 2778
XM_017003115.1 XP_016858604.1 5 359
GenBank mRNA AF006085.1 - 5 410
AF279893.1 - 4 252
AK092520.1 - 2 1457
AK093625.1 - 2 1448
AK290268.1 - 6 394
BC000590.2 - 6 358 3346685
BG282181.1 - 6 494
BG765898.1 - 6 438
BT006898.1 - 5 326
U50523.1 - 6 410

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

movement of cell or subcellular component
positive regulation of lamellipodium assembly
Arp2/3 complex-mediated actin nucleation
Fc-gamma receptor signaling pathway involved in phagocytosis
ephrin receptor signaling pathway
positive regulation of substrate adhesion-dependent cell spreading
structural constituent of cytoskeleton
protein binding
actin filament binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0