This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KRTAP5-11 SIRNA Assays ›
Gene Symbol:
KRTAP5-11
Gene Name:
keratin associated protein 5-11
Gene Aliases:
KRTAP5-5, KRTAP5-6, KRTAP5.11
Chromosome Location:
Chr.11: 71581855 - 71582875 on Build GRCh38
UniGene ID:
Hs.679617
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s196512

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: KRTAP5-11
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005405.2 NP_001005405.1 1 590
GenBank mRNA AB126080.1 - 1 590
AJ628244.1 - 1 12
BC130498.1 - 1 578
BC130502.1 - 1 578

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0