This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HUWE1 SIRNA Assays ›
Gene Symbol:
HUWE1
Gene Name:
HECT, UBA and WWE domain containing 1, E3 ubiquitin protein ligase
Gene Aliases:
ARF-BP1, HECTH9, HSPC272, Ib772, LASU1, MULE, URE-B1, UREB1
Chromosome Location:
Chr.X: 53532096 - 53686729 on Build GRCh38
UniGene ID:
Hs.136905
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s19596
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HUWE1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_031407.6
NP_113584.3
68
10491
XM_005261965.3
XP_005262022.1
67
10558
XM_011530747.1
XP_011529049.1
69
10734
XM_011530751.2
XP_011529053.1
68
10602
XM_017029191.1
XP_016884680.1
67
13486
XM_017029192.1
XP_016884681.1
67
13483
XM_017029193.1
XP_016884682.1
67
13465
XM_017029194.1
XP_016884683.1
67
13486
XM_017029195.1
XP_016884684.1
67
13438
XM_017029196.1
XP_016884685.1
67
13435
XM_017029197.1
XP_016884686.1
66
13387
XM_017029198.1
XP_016884687.1
68
10828
XM_017029199.1
XP_016884688.1
69
10755
XM_017029200.1
XP_016884689.1
68
10625
XM_017029201.1
XP_016884690.1
68
10642
XM_017029202.1
XP_016884691.1
68
10618
XM_017029203.1
XP_016884692.1
68
10624
XM_017029204.1
XP_016884693.1
66
13236
XM_017029205.1
XP_016884694.1
65
13114
XM_017029206.1
XP_016884695.1
64
13084
GenBank mRNA
AB002310.3
-
37
6542
AB071605.1
-
41
7191
AF161390.1
-
14
2738
AY772009.1
-
66
10136
AY929612.1
-
65
10088
DQ097177.1
-
68
10490
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_021523, BC011391, DQ097265, AY929611, AY772010, and others.
Gene Ontology Categories:
Function(s)
Process(es)
protein polyubiquitination
base-excision repair
protein monoubiquitination
histone ubiquitination
cell differentiation
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
positive regulation of protein targeting to mitochondrion
DNA binding
ubiquitin-protein transferase activity
protein binding
ligase activity
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0