This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CMTM5 SIRNA Assays ›
Gene Symbol:
CMTM5
Gene Name:
CKLF like MARVEL transmembrane domain containing 5
Gene Aliases:
CKLFSF5
Chromosome Location:
Chr.14: 23376439 - 23379772 on Build GRCh38
UniGene ID:
Hs.99272
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s195619

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CMTM5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037288.1 NP_001032365.1 4 1000
NM_001288744.1 NP_001275673.1 4 940
NM_001288745.1 NP_001275674.1 3 847
NM_001288746.1 NP_001275675.1 6 1294
NM_138460.2 NP_612469.1 5 1093
XM_017020953.1 XP_016876442.1 6 1079
XM_017020954.1 XP_016876443.1 6 956
GenBank mRNA AF479262.2 - 5 1094
AF527413.1 - 6 1077
AF527414.1 - 5 724
AF527948.2 - 3 618
AF527949.1 - 4 711
AK094840.1 - 4 1825
BC013109.1 - 5 838 4214683

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

chemotaxis
negative regulation of myoblast differentiation
cytokine activity
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0