This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ENDOG SIRNA Assays ›
Gene Symbol:
ENDOG
Gene Name:
endonuclease G
Gene Aliases:
-
Chromosome Location:
Chr.9: 128818402 - 128822676 on Build GRCh38
UniGene ID:
Hs.745161
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s194395
Validated

Purification
Size
Availability Inventoried
Catalog # 4390824
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ENDOG
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004435.2 NP_004426.2 2 756
XM_011518347.2 XP_011516649.1 2 854
GenBank mRNA AK130973.1 - 2 237
BC004922.2 - 2 631 3604703
BC016351.1 - 2 786 4080883
X79444.1 - 2 737

Back To Top

More Information


Matching Taqman Gene Expression Assay:


Hs00172770_m1

Gene Ontology Categories:

Function(s) Process(es)

in utero embryonic development
apoptotic DNA fragmentation
DNA recombination
aging
response to mechanical stimulus
response to estradiol
response to tumor necrosis factor
neuron death in response to oxidative stress
response to antibiotic
cellular response to calcium ion
cellular response to glucose stimulus
cellular response to hypoxia
positive regulation of hydrogen peroxide-mediated programmed cell death
positive regulation of apoptotic DNA fragmentation
nucleic acid binding
endonuclease activity
deoxyribonuclease activity
protein binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0