This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other C8G SIRNA Assays ›
Gene Symbol:
C8G
Gene Name:
complement component 8, gamma polypeptide
Gene Aliases:
C8C
Chromosome Location:
Chr.9: 136944870 - 136946981 on Build GRCh38
UniGene ID:
Hs.1285
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s194320

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: C8G
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000606.2 NP_000597.2 1 194
GenBank mRNA BC113624.1 - 1 169 8322676
BC113626.1 - 1 169 8322678
BM852158.1 - 1 194
M17263.1 - 1 145
M17999.1 - 1 180
X06465.1 - 1 180

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

complement activation, alternative pathway
complement activation, classical pathway
cytolysis
regulation of complement activation
complement binding
retinol binding
protein complex binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header