This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LOC100359498 SIRNA Assays ›
Gene Symbol:
LOC100359498 LOC103691938 Rpl35a
Gene Name:
ribosomal protein L35a-like
60S ribosomal protein L35a-like
ribosomal protein L35a
Gene Aliases:
-
-
-
Chromosome Location:
Chr.11: 70963223 - 70967164 on Build Rnor_6.0
Chromosome Location:
Chr.3: 149889546 - 149890041 on Build Rnor_6.0
Chromosome Location:
Chr.12: 7186459 - 7186874 on Build Rnor_6.0
UniGene ID:
Rn.201355
-
Rn.156479
Species:
Rat
Species Specific ID (Flybase ID):
-
-
-
siRNA ID s193950

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LOC100359498
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_002727895.4 XP_002727941.1 3 218
XM_002727896.4 XP_002727942.1 3 223
GenBank mRNA FQ210925.1 - 3 187
Gene Symbol: LOC103691938
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008762404.1 XP_008760626.1 2 132
Gene Symbol: Rpl35a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021264.3 NP_067087.1 127
GenBank mRNA BC061557.1 - 193 6917939
X03475.1 - 127

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

translation
tRNA binding
structural constituent of ribosome

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0