This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RNPC3 SIRNA Assays ›
Gene Symbol:
Rnpc3
Gene Name:
RNA-binding region (RNP1, RRM) containing 3
Gene Aliases:
-
Chromosome Location:
Chr.2: 216481457 - 216510054 on Build Rnor_6.0
UniGene ID:
Rn.17587
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s193872

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rnpc3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100810.1 NP_001094280.1 4 453
XM_006233211.2 XP_006233273.1 4 831
XM_008761424.1 XP_008759646.1 4 831
XM_008761425.1 XP_008759647.1 4 832
GenBank mRNA BC098742.1 - 2 131
BC166571.1 - 4 453 7320328

Back To Top

More Information


Gene Ontology Categories:

Process(es)

nucleotide binding
RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm