This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RPS18 SIRNA Assays ›
Gene Symbol:
Rps18 LOC100360679
Gene Name:
ribosomal protein S18
ribosomal protein S18-like
Gene Aliases:
Ke3
-
Chromosome Location:
Chr.20: 5441875 - 5445553 on Build Rnor_6.0
Chromosome Location:
Chr.10: 104521700 - 104522235 on Build Rnor_6.0
UniGene ID:
Rn.42766
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID s193521

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rps18
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_213557.1 NP_998722.1 3 298
GenBank mRNA BC126072.1 - 3 272 8362450
FQ209845.1 - 3 296
FQ210672.1 - 3 280
FQ210792.1 - 3 296
FQ217732.1 - 3 296
FQ220696.1 - 3 296
FQ221730.1 - 3 296
FQ221918.1 - 3 296
FQ221943.1 - 1 106
FQ222652.1 - 3 296
FQ222675.1 - 3 294
FQ223127.1 - 3 296
FQ223169.1 - 3 297
FQ223198.1 - 3 296
FQ223436.1 - 3 296
FQ223481.1 - 3 296
X57529.1 - 3 290
Gene Symbol: LOC100360679
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_002727833.4 XP_002727879.1 1 290

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_022551, BC071678, AY536376, BC106063, X69150, and others.

Gene Ontology Categories:

Function(s) Process(es)

translation
regulation of translation
ribosome biogenesis
structural constituent of ribosome
rRNA binding
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0