This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MAMSTR SIRNA Assays ›
Gene Symbol:
Mamstr
Gene Name:
MEF2 activating motif and SAP domain containing transcriptional regulator
Gene Aliases:
-
Chromosome Location:
Chr.1: 101621704 - 101628379 on Build Rnor_6.0
UniGene ID:
-
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s193464

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mamstr
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_001076631.5 XP_001076631.2 6 1469
XM_006229210.2 XP_006229272.1 6 1854
XM_006229211.2 XP_006229273.1 6 1413

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_172418, DQ534901, AK052976, AK036109, BC119481, and others.

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
positive regulation of myotube differentiation
positive regulation of transcription from RNA polymerase II promoter
transcription factor activity, RNA polymerase II transcription factor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0