This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other POLR2E SIRNA Assays ›
Gene Symbol:
Polr2e
Gene Name:
polymerase (RNA) II (DNA directed) polypeptide E
Gene Aliases:
-
Chromosome Location:
Chr.7: 12532746 - 12536797 on Build Rnor_6.0
UniGene ID:
Rn.65038
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s193414

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Polr2e
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109614.1 NP_001103084.1 3 455
XM_006241009.2 XP_006241071.1 3 584
XM_006241010.1 XP_006241072.1 3 475
XM_006241011.1 XP_006241073.1 3 502
GenBank mRNA BC158787.1 - 3 413 7304442
FQ219797.1 - 3 387
FQ220392.1 - 3 385
FQ229401.1 - 3 502
FQ230290.1 - 3 390
FQ234550.1 - 3 435

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription from RNA polymerase I promoter
transcription from RNA polymerase II promoter
transcription from RNA polymerase III promoter
biological_process
RNA polymerase I activity
RNA polymerase II activity
RNA polymerase III activity
molecular_function
DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0