This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TAS2R130 SIRNA Assays ›
Gene Symbol:
Tas2r130
Gene Name:
taste receptor, type 2, member 130
Gene Aliases:
Tas2r7
Chromosome Location:
Chr.4: 165746263 - 165747201 on Build Rnor_6.0
UniGene ID:
Rn.134167
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s192950

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tas2r130
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_001074122.3 XP_001074122.1 1 205
GenBank mRNA AF240766.1 - 1 160

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_199156, BC119329.

Gene Ontology Categories:

Function(s) Process(es)

detection of chemical stimulus involved in sensory perception of bitter taste
G-protein coupled receptor signaling pathway
biological_process
taste receptor activity
bitter taste receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0