This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SMARCAL1 SIRNA Assays ›
Gene Symbol:
Smarcal1
Gene Name:
Swi/SNF related matrix associated, actin dependent regulator of chromatin, subfamily a-like 1
Gene Aliases:
-
Chromosome Location:
Chr.9: 79943757 - 79990230 on Build Rnor_6.0
UniGene ID:
Rn.34679
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s192933

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Smarcal1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108222.1 NP_001101692.1 1 33
XM_006245212.2 XP_006245274.1 1 408
XM_008767236.1 XP_008765458.1 1 407

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

DNA strand renaturation
DNA metabolic process
DNA repair
regulation of transcription from RNA polymerase II promoter
cellular response to DNA damage stimulus
biological_process
chromatin modification
replication fork processing
molecular_function
DNA binding
helicase activity
ATP binding
DNA-dependent ATPase activity
annealing helicase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0