This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SEC16A SIRNA Assays ›
Gene Symbol:
SEC16A
Gene Name:
SEC16 homolog A, endoplasmic reticulum export factor
Gene Aliases:
KIAA0310, SEC16L, p250
Chromosome Location:
Chr.9: 136440096 - 136484552 on Build GRCh38
UniGene ID:
Hs.522500
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s19237
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SEC16A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001276418.1
NP_001263347.1
3
3905
NM_014866.1
NP_055681.1
4
3895
XM_005266134.2
XP_005266191.1
3
3758
XM_011519246.2
XP_011517548.1
4
3904
XM_011519247.1
XP_011517549.1
3
3761
XM_011519248.2
XP_011517550.1
3
3735
XM_011519249.1
XP_011517551.1
3
3758
XM_011519250.2
XP_011517552.1
3
3758
XM_011519251.1
XP_011517553.1
3
3758
XM_011519252.2
XP_011517554.1
3
3758
XM_011519253.1
XP_011517555.1
3
3758
XM_011519254.1
XP_011517556.1
3
3758
XM_011519255.2
XP_011517557.1
3
3758
XM_011519256.1
XP_011517558.1
3
3758
XM_011519257.1
XP_011517559.1
3
3758
XM_011519258.1
XP_011517560.1
3
3758
XM_011519259.1
XP_011517561.1
3
3758
XM_011519260.2
XP_011517562.1
3
3742
XM_011519261.1
XP_011517563.1
3
3758
XM_011519262.1
XP_011517564.1
4
3047
XM_011519263.2
XP_011517565.1
4
3047
XM_011519264.2
XP_011517566.1
3
3758
XM_017015363.1
XP_016870852.1
3
3758
XM_017015364.1
XP_016870853.1
3
3758
GenBank mRNA
AB002308.4
-
4
3895
AB593115.1
-
3
3905
BC125018.1
-
2
2467
BC125019.1
-
3
1756
DQ903855.1
-
2
3052
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
endoplasmic reticulum organization
protein transport
substantia nigra development
COPII vesicle coating
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0