This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TBC1D4 SIRNA Assays ›
Gene Symbol:
TBC1D4
Gene Name:
TBC1 domain family member 4
Gene Aliases:
AS160, NIDDM5
Chromosome Location:
Chr.13: 75284664 - 75482646 on Build GRCh38
UniGene ID:
Hs.210891
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s19141

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 357.00
Your Price
Online offer: 296.31
296.3
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TBC1D4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286658.2 NP_001273587.1 3 1496
NM_001286659.2 NP_001273588.1 3 1496
NM_014832.4 NP_055647.2 3 1496
XM_005266603.1 XP_005266660.1 3 1974
XM_005266605.2 XP_005266662.1 3 824
XM_006719903.3 XP_006719966.1 3 1184
XM_011535331.1 XP_011533633.1 3 1974
XM_017020882.1 XP_016876371.1 3 705
XM_017020883.1 XP_016876372.1 3 810
GenBank mRNA AB011175.1 - 3 1442
AB449885.1 - 3 1095
AK307493.1 - 3 1496
BC151239.1 - 3 1434 8860512
FM207106.1 - 3 1095
FM207107.1 - 3 1095

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

intracellular protein transport
vesicle-mediated transport
regulation of vesicle fusion
negative regulation of vesicle fusion
cellular response to insulin stimulus
membrane organization
activation of GTPase activity
GTPase activator activity
Rab GTPase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0