This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MEP1A SIRNA Assays ›
Gene Symbol:
Mep1a
Gene Name:
meprin 1 alpha
Gene Aliases:
-
Chromosome Location:
Chr.9: 20048124 - 20083429 on Build Rnor_6.0
UniGene ID:
Rn.16265
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s191286

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mep1a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013143.1 NP_037275.1 13 2063
XM_008766837.1 XP_008765059.1 13 2102
XM_008766838.1 XP_008765060.1 11 1924
GenBank mRNA BC081834.1 - 13 2078 7128404
S43408.1 - 13 2063

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_005588, M82962, AK225212, BC143650, AK315246, and others.

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
metalloendopeptidase activity
zinc ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0