This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MEIOB SIRNA Assays ›
Gene Symbol:
Meiob
Gene Name:
meiosis specific with OB domains
Gene Aliases:
-
Chromosome Location:
Chr.10: 14174645 - 14206026 on Build Rnor_6.0
UniGene ID:
Rn.200954
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s188056

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Meiob
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001115033.1 NP_001108505.1 13 1281
XM_006246031.2 XP_006246093.1 13 1496
GenBank mRNA BC098928.1 - 5 1145
BC158604.1 - 13 1281 7134815

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_029197, BC119085, BC119087, AK015924.

Gene Ontology Categories:

Function(s) Process(es)

resolution of meiotic recombination intermediates
double-strand break repair via homologous recombination
DNA catabolic process, exonucleolytic
synapsis
male meiosis
male meiosis I
female meiosis I
fertilization
chromatin binding
single-stranded DNA binding
single-stranded DNA 3'-5' exodeoxyribonuclease activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0