This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GALNT6 SIRNA Assays ›
Gene Symbol:
Galnt6
Gene Name:
polypeptide N-acetylgalactosaminyltransferase 6
Gene Aliases:
-
Chromosome Location:
Chr.7: 142322172 - 142364283 on Build Rnor_6.0
UniGene ID:
Rn.104292
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s186027

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Galnt6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001172063.2 NP_001165534.1 10 1792
XM_006242294.2 XP_006242356.1 12 2878
XM_006242295.2 XP_006242357.1 12 2201
XM_006242296.2 XP_006242358.1 12 2145
XM_008765651.1 XP_008763873.1 13 2380
XM_008765652.1 XP_008763874.1 12 2137
XM_008765653.1 XP_008763875.1 12 2417
GenBank mRNA FQ211776.1 - 12 2169

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein glycosylation
polypeptide N-acetylgalactosaminyltransferase activity
carbohydrate binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0