This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FUBP1 SIRNA Assays ›
Gene Symbol:
Fubp1
Gene Name:
far upstream element (FUSE) binding protein 1
Gene Aliases:
-
Chromosome Location:
Chr.2: 257425676 - 257452745 on Build Rnor_6.0
UniGene ID:
Rn.177078
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s181850

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fubp1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037653.1 NP_001032742.1 17 1727
XM_006233507.2 XP_006233569.1 18 1829
XM_006233511.2 XP_006233573.1 18 1829
XM_006233512.2 XP_006233574.1 18 1829
XM_006233513.2 XP_006233575.1 17 1765
XM_008761553.1 XP_008759775.1 18 1843
XM_008761554.1 XP_008759776.1 18 1829
XM_008761555.1 XP_008759777.1 17 1765
XM_008761556.1 XP_008759778.1 17 1762
XM_008761557.1 XP_008759779.1 17 1762
GenBank mRNA BC107942.1 - 17 1727 7389449

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
transcription from RNA polymerase II promoter
positive regulation of gene expression
positive regulation of transcription from RNA polymerase II promoter
transcriptional activator activity, RNA polymerase II distal enhancer sequence-specific binding
DNA binding
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0