This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MED24 SIRNA Assays ›
Gene Symbol:
Med24
Gene Name:
mediator complex subunit 24
Gene Aliases:
Thrap4
Chromosome Location:
Chr.10: 86620166 - 86645564 on Build Rnor_6.0
UniGene ID:
Rn.7360
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s181655

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Med24
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034079.1 NP_001029251.1 2 149
XM_006247415.2 XP_006247477.1 2 189
XM_006247416.2 XP_006247478.1 2 235
GenBank mRNA BC097461.1 - 2 149 7457533
FQ210692.1 - 2 172

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011869, AK192144, AK158385, AK193218, AF126543, and others.

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
protein ubiquitination
histone acetylation
stem cell population maintenance
intracellular steroid hormone receptor signaling pathway
androgen receptor signaling pathway
positive regulation of transcription, DNA-templated
protein heterooligomerization
RNA polymerase II transcription cofactor activity
transcription cofactor activity
transcription coactivator activity
histone acetyltransferase activity
receptor activity
thyroid hormone receptor binding
ubiquitin protein ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0