This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SURF2 SIRNA Assays ›
Gene Symbol:
Surf2 LOC100912042
Gene Name:
surfeit 2
surfeit locus protein 2-like
Gene Aliases:
-
-
Chromosome Location:
Chr.3: 5464593 - 5468426 on Build Rnor_6.0
Chromosome Location:
Chr.3: 4872666 - 4876499 on Build Rnor_6.0
UniGene ID:
Rn.162644
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID s181634

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Surf2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033866.1 NP_001029038.1 587
XM_006233850.1 XP_006233912.1 598
GenBank mRNA BC059157.1 - 586 6918490
CO384344.1 - 587
FQ219299.1 - 613
FQ229667.1 - 562
Gene Symbol: LOC100912042
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003749433.2 XP_003749481.1 5 598
XM_006233723.1 XP_006233785.1 5 598

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0