This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RGD1564955 SIRNA Assays ›
Gene Symbol:
RGD1564955
Gene Name:
similar to fibrous sheath interacting protein 2
Gene Aliases:
-
Chromosome Location:
Chr.X: 135729374 - 135761064 on Build Rnor_6.0
UniGene ID:
Rn.127617
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s180535

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: RGD1564955
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006257545.1 XP_006257607.1 2 213
XM_006257547.2 XP_006257609.1 2 212
XM_008773543.1 XP_008771765.1 2 212
XM_008773544.1 XP_008771766.1 2 212
XM_008773545.1 XP_008771767.1 2 256
GenBank mRNA BC160914.1 - 2 219

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0