This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RGD1562161 SIRNA Assays ›
Gene Symbol:
RGD1562161
Gene Name:
similar to chromosome X open reading frame 23
Gene Aliases:
-
Chromosome Location:
Chr.X: 38294742 - 38353517 on Build Rnor_6.0
UniGene ID:
Rn.214618
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s180456

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: RGD1562161
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127568.1 NP_001121040.1 8 1794
XM_006256933.2 XP_006256995.1 8 1849
XM_006256934.2 XP_006256996.1 8 2125
XM_006256936.2 XP_006256998.1 8 1850
XM_006256937.2 XP_006256999.1 8 1849
XM_006256938.2 XP_006257000.1 8 1849
XM_008773186.1 XP_008771408.1 8 1849
GenBank mRNA BC166438.1 - 8 1794 7441789

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001033472, AK145159, AK044001.

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0