siRNA Details
Gene Symbol: Ppip5k2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025775.1 | NP_001020946.1 | 13 | 1640 | ||
GenBank mRNA | BC097365.1 | - | 13 | 1640 | 7443073 |

Gene Symbol: Ppip5k2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025775.1 | NP_001020946.1 | 13 | 1640 | ||
GenBank mRNA | BC097365.1 | - | 13 | 1640 | 7443073 |