This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SATB2 SIRNA Assays ›
Gene Symbol:
Satb2
Gene Name:
SATB homeobox 2
Gene Aliases:
RGD1562369
Chromosome Location:
Chr.9: 63456877 - 63641408 on Build Rnor_6.0
UniGene ID:
Rn.213596
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s180183

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Satb2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109306.1 NP_001102776.1 6 963
XM_006244931.2 XP_006244993.1 6 1635
XM_008767128.1 XP_008765350.1 5 1407
XM_008767129.1 XP_008765351.1 6 909

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_139146, AJ606045, BC138626, BC138625, AF319623, and others.

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
neuron migration
osteoblast development
chromatin remodeling
transcription, DNA-templated
embryonic pattern specification
regulation of gene expression
commitment of neuronal cell to specific neuron type in forebrain
positive regulation of transcription from RNA polymerase II promoter
embryonic skeletal system morphogenesis
cartilage development
palate development
cellular response to organic substance
DNA binding
chromatin binding
sequence-specific DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0