This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RGD1559960 SIRNA Assays ›
Gene Symbol:
RGD1559960 Sult1c2a
Gene Name:
similar to Sulfotransferase K1 (rSULT1C2)
sulfotransferase family, cytosolic, 1C, member 2a
Gene Aliases:
-
ST1C2A, Sult1c1
Chromosome Location:
Chr.9: 4867054 - 4879861 on Build Rnor_6.0
Chromosome Location:
Chr.9: 4152588 - 4168352 on Build Rnor_6.0
UniGene ID:
-
Rn.205998
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID s180101

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: RGD1559960
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006244256.2 XP_006244318.2 4 546
XM_008766792.1 XP_008765014.1 5 693
Gene Symbol: Sult1c2a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013177.2 NP_001013195.2 862
GenBank mRNA AJ238392.1 - 803
BC081691.1 - 759 7110136
BC092564.1 - 862 7372924
FQ209913.1 - 856

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

metabolic process
sulfotransferase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0