This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZFP259 SIRNA Assays ›
Gene Symbol:
Zfp259
Gene Name:
zinc finger protein 259
Gene Aliases:
RGD1562173
Chromosome Location:
Chr.8: 50563047 - 50572621 on Build Rnor_6.0
UniGene ID:
Rn.102011
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s179982

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Zfp259
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001137646.1 NP_001131118.1 2 345

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

microtubule cytoskeleton organization
inner cell mass cell proliferation
trophectodermal cell proliferation
cell proliferation
positive regulation of gene expression
spinal cord development
Cajal body organization
regulation of myelination
positive regulation of RNA splicing
DNA endoreduplication
positive regulation of protein import into nucleus
positive regulation of growth
axon development
cellular response to epidermal growth factor stimulus
positive regulation of transcription involved in G1/S transition of mitotic cell cycle
apoptotic process involved in development
pre-mRNA catabolic process
negative regulation of motor neuron apoptotic process
zinc ion binding
receptor tyrosine kinase binding
translation initiation factor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header