This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LOC500827 SIRNA Assays ›
Gene Symbol:
LOC500827
Gene Name:
similar to hypothetical protein FLJ35821
Gene Aliases:
-
Chromosome Location:
Chr.7: 40316154 - 40350482 on Build Rnor_6.0
UniGene ID:
Rn.123316
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s179759

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LOC500827
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024352.1 NP_001019523.2 11 1480
XM_006241290.2 XP_006241352.1 11 1346
XM_006241291.2 XP_006241353.1 12 1978
XM_008765319.1 XP_008763541.1 11 1999
GenBank mRNA BC078911.1 - 11 1480 7112362

Back To Top

More Information


Gene Ontology Categories:

Process(es)

metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0