This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other EFCAB11 SIRNA Assays ›
Gene Symbol:
Efcab11
Gene Name:
EF-hand calcium binding domain 11
Gene Aliases:
-
Chromosome Location:
Chr.6: 123742971 - 123894816 on Build Rnor_6.0
UniGene ID:
Rn.119809
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s179658

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Efcab11
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024350.1 NP_001019521.1 4 363
XM_006240446.2 XP_006240508.2 4 465
GenBank mRNA BC079408.1 - 4 363 7113005

Back To Top

More Information


Gene Ontology Categories:

Process(es)

calcium ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0