This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NRXN2 SIRNA Assays ›
Gene Symbol:
NRXN2
Gene Name:
neurexin 2
Gene Aliases:
-
Chromosome Location:
Chr.11: 64606174 - 64723188 on Build GRCh38
UniGene ID:
Hs.372938
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s17950
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NRXN2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_015080.3
NP_055895.1
23
5003
NM_138732.2
NP_620060.1
20
4793
NM_138734.2
NP_620063.1
7
1915
XM_005274400.3
XP_005274457.1
22
4535
XM_011545370.1
XP_011543672.1
22
4565
XM_011545371.1
XP_011543673.1
22
4562
XM_011545373.1
XP_011543675.1
22
4544
XM_011545375.1
XP_011543677.1
21
4520
XM_011545385.1
XP_011543687.1
15
2910
XM_017018563.1
XP_016874052.1
21
4535
XM_017018564.1
XP_016874053.1
21
4532
XM_017018565.1
XP_016874054.1
21
4529
XM_017018566.1
XP_016874055.1
21
4493
XM_017018567.1
XP_016874056.1
21
4475
XM_017018568.1
XP_016874057.1
21
4466
XM_017018569.1
XP_016874058.1
20
4463
XM_017018572.1
XP_016874061.1
22
3860
XM_017018574.1
XP_016874063.1
21
3830
XM_017018575.1
XP_016874064.1
21
3827
XM_017018576.1
XP_016874065.1
17
3475
XM_017018577.1
XP_016874066.1
15
2901
GenBank mRNA
AB023138.2
-
21
4770
AB035266.1
-
22
4655
AB177862.1
-
19
3707
AK098211.1
-
3
1612
BC150275.1
-
21
4770
9007249
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
neuron cell-cell adhesion
signal transduction
chemical synaptic transmission
neurotransmitter secretion
synapse assembly
adult behavior
social behavior
vocal learning
vocalization behavior
postsynaptic membrane assembly
gephyrin clustering involved in postsynaptic density assembly
neuroligin clustering involved in postsynaptic membrane assembly
postsynaptic density protein 95 clustering
transmembrane signaling receptor activity
calcium channel regulator activity
metal ion binding
cell adhesion molecule binding
neuroligin family protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0