siRNA Details
Gene Symbol: Ppp1r3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109222.1 | NP_001102692.1 | 4 | 2837 | ||
XM_008762717.1 | XP_008760939.1 | 4 | 2879 |

Gene Symbol: Ppp1r3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109222.1 | NP_001102692.1 | 4 | 2837 | ||
XM_008762717.1 | XP_008760939.1 | 4 | 2879 |