This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ASB4 SIRNA Assays ›
Gene Symbol:
Asb4
Gene Name:
ankyrin repeat and SOCS box-containing 4
Gene Aliases:
Asb-4
Chromosome Location:
Chr.4: 30461686 - 30501340 on Build Rnor_6.0
UniGene ID:
Rn.162732
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s178787

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Asb4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024318.1 NP_001019489.1 4 1024
XM_008762714.1 XP_008760936.1 5 1430
GenBank mRNA AY577765.1 - 4 1024

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

intracellular signal transduction
protein autoubiquitination
positive regulation of vasculogenesis
ubiquitin-protein transferase activity
enzyme binding
ubiquitin protein ligase binding
ubiquitin protein ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0