This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PRPF38B SIRNA Assays ›
Gene Symbol:
Prpf38b
Gene Name:
pre-mRNA processing factor 38B
Gene Aliases:
-
Chromosome Location:
Chr.2: 211706245 - 211715271 on Build Rnor_6.0
UniGene ID:
Rn.7210
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s178351

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Prpf38b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024305.2 NP_001019476.1 5 967
XM_006233172.1 XP_006233234.1 6 723
XM_008761421.1 XP_008759643.1 5 1018
GenBank mRNA BC079264.1 - 5 916 7133560
FQ210223.1 - 5 947

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

mRNA processing
RNA splicing
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0