This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LEKR1 SIRNA Assays ›
Gene Symbol:
Lekr1
Gene Name:
leucine, glutamate and lysine rich 1
Gene Aliases:
RGD1562738, RGD1565425
Chromosome Location:
Chr.2: 157452004 - 157621908 on Build Rnor_6.0
UniGene ID:
Rn.198594
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s178282

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Lekr1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001115029.2 NP_001108501.2 9 1404
XM_006232472.2 XP_006232534.1 8 2219
XM_006232473.1 XP_006232535.1 9 1066
XM_006232474.2 XP_006232536.1 8 1278
XM_006232475.2 XP_006232537.1 8 929

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001166659.

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0