This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNHIT3 SIRNA Assays ›
Gene Symbol:
ZNHIT3
Gene Name:
zinc finger HIT-type containing 3
Gene Aliases:
TRIP3
Chromosome Location:
Chr.17: 36486627 - 36499310 on Build GRCh38
UniGene ID:
Hs.2210
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s17823

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNHIT3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001281432.1 NP_001268361.1 3 210
NM_001281433.1 NP_001268362.1 3 210
NM_004773.3 NP_004764.1 3 210
NR_104009.1 3 205
NR_104010.1 2 178
NR_104011.1 2 178
GenBank mRNA AB209193.1 - 1 3674
AF400652.1 - 3 202
AK290858.1 - 3 161
AK308583.1 - 2 106
AK310022.1 - 3 145
AV763423.1 - 177
BC017931.1 - 3 175 4669658
BC105608.1 - 2 150
BG032116.1 - 3 145
BI767446.1 - 3 143
BM457955.1 - 140
CB996852.1 - 3 235
HY045632.1 - 3 211
L40410.1 - 3 128

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
protein binding
metal ion binding
thyroid hormone receptor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0