This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DDIAS SIRNA Assays ›
Gene Symbol:
Ddias
Gene Name:
DNA damage-induced apoptosis suppressor
Gene Aliases:
RGD1559690, noxin
Chromosome Location:
Chr.1: 157675754 - 157700064 on Build Rnor_6.0
UniGene ID:
Rn.14573
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s177803

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ddias
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001126294.1 NP_001119766.1 6 1458
GenBank mRNA BC166470.1 - 6 1458 9088340

Back To Top

More Information


Gene Ontology Categories:

Function(s)

cellular response to DNA damage stimulus
spermatid development
hemopoiesis
cellular response to UV
cellular response to cytokine stimulus
cellular response to hydroperoxide
cellular response to gamma radiation
mitotic cell cycle arrest
negative regulation of fibroblast apoptotic process

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0