This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FBXO31 SIRNA Assays ›
Gene Symbol:
Fbxo31
Gene Name:
F-box protein 31
Gene Aliases:
RGD1561069
Chromosome Location:
Chr.19: 53487616 - 53625673 on Build Rnor_6.0
UniGene ID:
Rn.163412
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s177521

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fbxo31
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001044259.2 NP_001037724.2 5 655
XM_006255719.2 XP_006255781.1 5 1649
XM_006255720.2 XP_006255782.1 4 1562
GenBank mRNA BC110046.1 - 3 761 7302542
BC166840.1 - 5 655 9027777

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_133765, AK009859, AK140412, AK163641, AK197038, and others.

Gene Ontology Categories:

Function(s) Process(es)

cellular response to DNA damage stimulus
negative regulation of cyclin-dependent protein serine/threonine kinase by cyclin degradation
protein ubiquitination
SCF-dependent proteasomal ubiquitin-dependent protein catabolic process
mitotic G1 DNA damage checkpoint
cyclin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0