This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FAM170A SIRNA Assays ›
Gene Symbol:
Fam170a
Gene Name:
family with sequence similarity 170, member A
Gene Aliases:
RGD1565393
Chromosome Location:
Chr.18: 45023938 - 45028670 on Build Rnor_6.0
UniGene ID:
Rn.231132
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s177401

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fam170a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001126293.1 NP_001119765.1 2 620
XM_006254732.1 XP_006254794.1 4 1220
XM_006254733.2 XP_006254795.1 4 1214
XM_006254734.2 XP_006254796.1 4 1205
XM_008772162.1 XP_008770384.1 3 813
GenBank mRNA BC160906.1 - 4 1206 7455403

Back To Top

More Information


Gene Ontology Categories:

Function(s)

transcription from RNA polymerase II promoter
positive regulation of transcription, DNA-templated

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0