This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TCEA1 SIRNA Assays ›
Gene Symbol:
Tcea1 LOC498453
Gene Name:
transcription elongation factor A (SII) 1
similar to transcription elongation factor A 1 isoform 2
Gene Aliases:
-
-
Chromosome Location:
Chr.5: 14511183 - 14548490 on Build Rnor_6.0
Chromosome Location:
Chr.15: 11649873 - 11652109 on Build Rnor_6.0
UniGene ID:
Rn.6738
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID s176934

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tcea1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025735.1 NP_001020906.1 5 442
XM_006237816.2 XP_006237878.1 5 503
XM_008763546.1 XP_008761768.1 5 330
GenBank mRNA BC099141.1 - 5 442 7460791
FQ227501.1 - 5 414
Gene Symbol: LOC498453
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006251720.2 XP_006251782.1 1 496

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_201437, NM_006756, X62585, AK290454, AK026617, and others.
This siRNA also targets the following transcripts in Mouse: NM_001159751, NM_001159750, NM_011541, BC083127, AK136035, and others.

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
erythrocyte differentiation
regulation of DNA-templated transcription, elongation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of exoribonuclease activity
molecular_function
DNA binding
zinc ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0