This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other EIF2D SIRNA Assays ›
Gene Symbol:
Eif2d
Gene Name:
eukaryotic translation initiation factor 2D
Gene Aliases:
Lgtn
Chromosome Location:
Chr.13: 47935139 - 47958459 on Build Rnor_6.0
UniGene ID:
Rn.162387
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s176616

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Eif2d
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017489.1 NP_001017489.1 6 816
XM_006249790.2 XP_006249852.1 6 840
GenBank mRNA BC087701.1 - 6 816 7314085

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

formation of translation preinitiation complex
IRES-dependent translational initiation
ribosome disassembly
translation initiation factor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0