This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SLC8B1 SIRNA Assays ›
Gene Symbol:
Slc8b1
Gene Name:
solute carrier family 8 (sodium/lithium/calcium exchanger), member B1
Gene Aliases:
Slc24a6
Chromosome Location:
Chr.12: 41566980 - 41590287 on Build Rnor_6.0
UniGene ID:
Rn.13225
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s176574

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Slc8b1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017488.1 NP_001017488.1 8, 9 944
XM_008769249.1 XP_008767471.1 8, 9 987
GenBank mRNA BC079350.1 - 8, 9 944 7107820

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

mitochondrial calcium ion transport
biological_process
sodium ion transmembrane transport
glucose homeostasis
regulation of insulin secretion
regulation of cytosolic calcium ion concentration
mitochondrial calcium ion homeostasis
calcium ion transmembrane transport
calcium:sodium antiporter activity
identical protein binding
protein homodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0