This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other OLR545 SIRNA Assays ›
Gene Symbol:
Olr545 LOC103691842
Gene Name:
olfactory receptor 545
olfactory receptor 10AG1-like
Gene Aliases:
-
-
Chromosome Location:
Chr.3: 74906024 - 74906989 on Build Rnor_6.0
Chromosome Location:
Chr.3: 75167456 - 75168436 on Build Rnor_6.0
UniGene ID:
Rn.142914
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID s174982

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Olr545
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001053.1 NP_001001053.1 855
Gene Symbol: LOC103691842
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008762001.1 XP_008760223.1 1 870

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
detection of chemical stimulus involved in sensory perception of smell
olfactory receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0