This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other OLR774 SIRNA Assays ›
Gene Symbol:
Olr774 Olr776 Olr777
Gene Name:
olfactory receptor 774
olfactory receptor 776
olfactory receptor 777
Gene Aliases:
-
-
ratchr3-97368944-97368006_ORF
Chromosome Location:
Chr.3: 102995376 - 102996314 on Build Rnor_6.0
Chromosome Location:
Chr.3: 103065454 - 103066392 on Build Rnor_6.0
Chromosome Location:
Chr.3: 103089452 - 103090390 on Build Rnor_6.0
UniGene ID:
Rn.142756
Rn.142754
Rn.138179
Species:
Rat
Species Specific ID (Flybase ID):
-
-
-
siRNA ID s174596

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Olr774
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000918.1 NP_001000918.1 1 307
Gene Symbol: Olr776
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000608.1 NP_001000608.1 1 307
Gene Symbol: Olr777
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000579.1 NP_001000579.1 1 307

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
detection of chemical stimulus involved in sensory perception
detection of chemical stimulus involved in sensory perception of smell
transmembrane signaling receptor activity
G-protein coupled receptor activity
olfactory receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0