This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GAA SIRNA Assays ›
Gene Symbol:
Gaa
Gene Name:
glucosidase, alpha, acid
Gene Aliases:
-
Chromosome Location:
Chr.10: 108395874 - 108412999 on Build Rnor_6.0
UniGene ID:
Rn.162368
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s173326

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gaa
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199118.1 NP_954549.1 8 1485
XM_006247905.1 XP_006247967.1 8 1465
GenBank mRNA BC061753.1 - 8 1485 5599964

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of the force of heart contraction
diaphragm contraction
heart morphogenesis
glycogen metabolic process
glycogen catabolic process
striated muscle contraction
lysosome organization
locomotory behavior
tissue development
vacuolar sequestering
muscle cell cellular homeostasis
neuromuscular process controlling posture
neuromuscular process controlling balance
cardiac muscle contraction
alpha-1,4-glucosidase activity
carbohydrate binding
maltose alpha-glucosidase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0