This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LRRFIP1 SIRNA Assays ›
Gene Symbol:
Lrrfip1
Gene Name:
leucine rich repeat (in FLII) interacting protein 1
Gene Aliases:
-
Chromosome Location:
Chr.9: 98139485 - 98268148 on Build Rnor_6.0
UniGene ID:
Rn.153995
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s173269

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Lrrfip1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014269.1 NP_001014291.1 6, 7 735
XM_006245450.2 XP_006245512.1 18, 19 1338
XM_006245452.2 XP_006245514.1 17, 18 1266
XM_006245458.2 XP_006245520.1 6, 7 670
XM_006245462.2 XP_006245524.1 18, 19 1336
XM_006245463.2 XP_006245525.1 17, 18 1264
XM_006245464.2 XP_006245526.1 7, 8 741
XM_006245465.2 XP_006245527.1 6, 7 668
GenBank mRNA BC081883.1 - 6, 7 735 7108398

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
protein homodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0