This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other AZIN2 SIRNA Assays ›
Gene Symbol:
Azin2
Gene Name:
antizyme inhibitor 2
Gene Aliases:
Adc, AzI2, ODC-p, RGD1564776
Chromosome Location:
Chr.5: 147120335 - 147147615 on Build Rnor_6.0
UniGene ID:
Rn.50921
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s172781

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Azin2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC078981.1 - 1298 7112100
HQ413774.1 - 8 1147

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_052998, AB075825, AK095127, AK315677, AK122697, and others.
This siRNA also targets the following transcripts in Mouse: NM_172875, AK077201, BC138973, BC138974, AK154564, and others.

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0