This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PRPF6 SIRNA Assays ›
Gene Symbol:
Prpf6
Gene Name:
pre-mRNA processing factor 6
Gene Aliases:
RGD1307103
Chromosome Location:
Chr.3: 177098137 - 177162937 on Build Rnor_6.0
UniGene ID:
Rn.17062
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s172687

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Prpf6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001079766.1 NP_001073234.1 2 338
GenBank mRNA BC128779.1 - 2 338 7191091
FQ221925.1 - 2 341

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

spliceosomal tri-snRNP complex assembly
mRNA splicing, via spliceosome
RNA localization
positive regulation of transcription from RNA polymerase II promoter
transcription coactivator activity
RNA binding
ribonucleoprotein complex binding
poly(A) RNA binding
androgen receptor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0